missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Aminophenyl)carbazole hydrochloride, 98%

Catalog No. p-7045620
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11362MD 250 mg
AAL1136203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL11362MD Supplier Thermo Scientific Chemicals Supplier No. L11362MD
Only null left

CAS: 312700-07-5 | C18H15ClN2 | 294.782 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 312700-07-5
Molecular Formula C18H15ClN2
Molecular Weight (g/mol) 294.782
MDL Number MFCD00082678
InChI Key CFQROJKMLJRSAL-UHFFFAOYSA-N
Synonym 4-9h-carbazol-9-yl aniline hydrochloride, 9-4-aminophenyl carbazole hydrochloride, 4-carbazol-9-yl aniline hydrochloride, 4-carbazol-9-ylaniline hydrochloride, acmc-1agoy
PubChem CID 2794996
IUPAC Name 4-carbazol-9-ylaniline;hydrochloride
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)N.Cl

Specifications

Melting Point ∽250°C (decomposition)
Quantity 250 mg
Formula Weight 294.79
Percent Purity 98%
Chemical Name or Material 9-(4-Aminophenyl)carbazole hydrochloride
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only