missing translation for 'onlineSavingsMsg'
Learn More

9-(3-Bromophenyl)-9H-fluoren-9-ol 98.0+%, TCI America™

Catalog No. p-7124140
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B56151G 1 g
B56155G 5 g
2 options

Catalog No. B56151G

Supplier: TCI America B56151G

Only null left

Chemical Identifiers

CAS 1086641-47-5
Molecular Formula C19H13BrO
Molecular Weight (g/mol) 337.216
MDL Number MFCD16659044
InChI Key UKGUBHXRUOJAMM-UHFFFAOYSA-N
Synonym 9-(3-Bromophenyl)-9-hydroxy-9H-fluorene
PubChem CID 59404146
IUPAC Name 9-(3-bromophenyl)fluoren-9-ol
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC(=CC=C4)Br)O

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 1 g
Formula Weight 337.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(3-Bromophenyl)-9H-fluoren-9-ol