accessibility menu, dialog, popup

UserName

BrOH

CAS RN 1086641-47-5

IUPAC Name: 9-(3-bromophenyl)-9H-fluoren-9-ol
Synonyms: 9-(3-bromophenyl)fluoren-9-ol
Molecular Weight (g/mol): 337.22
Molecular Formula: C19H13BrO
InChi Key: UKGUBHXRUOJAMM-UHFFFAOYSA-N
SMILES: OC1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(Br)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results