missing translation for 'onlineSavingsMsg'
Learn More

9-(3-Bromophenyl)-9H-fluoren-9-ol 98.0+%, TCI America™

Catalog No. B56155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56155G 5 g
B56151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56155G Supplier TCI America Supplier No. B56155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1086641-47-5
Molecular Formula C19H13BrO
Molecular Weight (g/mol) 337.216
MDL Number MFCD16659044
InChI Key UKGUBHXRUOJAMM-UHFFFAOYSA-N
Synonym 9-(3-Bromophenyl)-9-hydroxy-9H-fluorene
PubChem CID 59404146
IUPAC Name 9-(3-bromophenyl)fluoren-9-ol
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC(=CC=C4)Br)O

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 5 g
Formula Weight 337.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(3-Bromophenyl)-9H-fluoren-9-ol