missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(2-Hydroxyethyl)anthracene 98.0+%, TCI America™
$512.10 - $1586.34
Chemical Identifiers
| CAS | 54060-73-0 |
|---|---|
| Molecular Formula | C16H14O |
| Molecular Weight (g/mol) | 222.287 |
| MDL Number | MFCD01318099 |
| InChI Key | KLNMQYHQWUWCPG-UHFFFAOYSA-N |
| Synonym | 2-(9-Anthryl)ethanol |
| PubChem CID | 281265 |
| IUPAC Name | 2-anthracen-9-ylethanol |
| SMILES | C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCO |
Chemical Identifiers
| 54060-73-0 | |
| 222.287 | |
| KLNMQYHQWUWCPG-UHFFFAOYSA-N | |
| 281265 | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCO |
| C16H14O | |
| MFCD01318099 | |
| 2-(9-Anthryl)ethanol | |
| 2-anthracen-9-ylethanol |
Specifications
| 54060-73-0 | |
| C16H14O | |
| 1 g | |
| KLNMQYHQWUWCPG-UHFFFAOYSA-N | |
| 2-anthracen-9-ylethanol | |
| 281265 | |
| ≥98.0% (GC) | |
| 9-(2-Hydroxyethyl)anthracene |
| 125°C | |
| MFCD01318099 | |
| 2-(9-Anthryl)ethanol | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCO | |
| 222.287 | |
| 222.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : No