accessibility menu, dialog, popup

UserName

CAS RN 54060-73-0

OH

CAS RN 54060-73-0

IUPAC Name: 2-(anthracen-9-yl)ethan-1-ol
Synonyms: 2-(anthracen-9-yl)ethanol
Molecular Weight (g/mol): 222.29
Molecular Formula: C16H14O
InChi Key: KLNMQYHQWUWCPG-UHFFFAOYSA-N
SMILES: OCCC1=C2C=CC=CC2=CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results