missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Dihydrophenanthrene 96.0+%, TCI America™
Supplier: TCI America D05545G
Specifications
| 9,10-Dihydrophenanthrene | |
| White-Yellow | |
| C14H12 | |
| 5 g | |
| XXPBFNVKTVJZKF-UHFFFAOYSA-N | |
| 9,10-dihydrophenanthrene | |
| 13058 | |
| ≥96.0% (GC) |
| 776-35-2 | |
| 314°C | |
| MFCD00001164 | |
| phenanthrene, 9,10-dihydro, unii-brm9tu2f34, 9,10-dihydro-phenanthrene, dihydrophenanthrene, brm9tu2f34, acmc-1ba4q, phenanthrene,9,10-dihydro, 9,10-dihydrophenanthrene, phenanthrene, 9,10-dihydro-8ci 9ci | |
| C1CC2=CC=CC=C2C3=CC=CC=C31 | |
| 180.25 | |
| 180.25 | |
| Crystalline Lumps |
Chemical Identifiers
| 776-35-2 | |
| 180.25 | |
| XXPBFNVKTVJZKF-UHFFFAOYSA-N | |
| 13058 | |
| C1CC2=CC=CC=C2C3=CC=CC=C31 |
| C14H12 | |
| MFCD00001164 | |
| phenanthrene, 9,10-dihydro, unii-brm9tu2f34, 9,10-dihydro-phenanthrene, dihydrophenanthrene, brm9tu2f34, acmc-1ba4q, phenanthrene,9,10-dihydro, 9,10-dihydrophenanthrene, phenanthrene, 9,10-dihydro-8ci 9ci | |
| 9,10-dihydrophenanthrene |
Safety and Handling
TSCA : Yes