missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dihydrophenanthrene 96.0+%, TCI America™

Catalog No. p-7126282
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05545G 5 g
D055425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05545G Supplier TCI America Supplier No. D05545G
Only null left

Chemical Identifiers

CAS 776-35-2
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00001164
InChI Key XXPBFNVKTVJZKF-UHFFFAOYSA-N
Synonym phenanthrene, 9,10-dihydro, unii-brm9tu2f34, 9,10-dihydro-phenanthrene, dihydrophenanthrene, brm9tu2f34, acmc-1ba4q, phenanthrene,9,10-dihydro, 9,10-dihydrophenanthrene, phenanthrene, 9,10-dihydro-8ci 9ci
PubChem CID 13058
IUPAC Name 9,10-dihydrophenanthrene
SMILES C1CC2=CC=CC=C2C3=CC=CC=C31

Specifications

Color White-Yellow
Boiling Point 314°C
Quantity 5 g
Formula Weight 180.25
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 9,10-Dihydrophenanthrene