missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dihydro-9,10-[3,4]thiophenoanthracene 98.0+%, TCI America™

Catalog No. D4412200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4412200MG 200 mg
D44121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4412200MG Supplier TCI America Supplier No. D4412200MG
Only null left

Chemical Identifiers

CAS 42490-26-6
Molecular Formula C18H12S
Molecular Weight (g/mol) 260.354
InChI Key GFAFZYPBTKDJBD-UHFFFAOYSA-N
PubChem CID 71206148
SMILES C1=CC=C2C3C4=CC=CC=C4C(C2=C1)C5=CSC=C35

Specifications

Melting Point 268°C
Color White-Yellow
Quantity 200 mg
Formula Weight 260.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dihydro-9,10-[3,4]thiophenoanthracene