accessibility menu, dialog, popup

UserName

S

CAS RN 42490-26-6

IUPAC Name: 17-thiapentacyclo[6.6.5.0²,⁷.0⁹,¹⁴.0¹⁵,¹⁹]nonadeca-2,4,6,9,11,13,15,18-octaene
Synonyms: 17-thiapentacyclo[6.6.5.0²,⁷.0⁹,¹⁴.0¹⁵,¹⁹]nonadeca-2,4,6,9,11,13,15,18-octaene
Molecular Weight (g/mol): 260.35
Molecular Formula: C18H12S
InChi Key: GFAFZYPBTKDJBD-UHFFFAOYSA-N
SMILES: S1C=C2C3C4=CC=CC=C4C(C2=C1)C1=CC=CC=C31
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results