missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dibromoanthracene 98.0+%, TCI America™

Catalog No. D0166500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0166500G 500 g
D016625G 25 g
D0166100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0166500G Supplier TCI America Supplier No. D0166500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 523-27-3
Molecular Formula C14H8Br2
Molecular Weight (g/mol) 336.026
MDL Number MFCD00001244
InChI Key BRUOAURMAFDGLP-UHFFFAOYSA-N
Synonym anthracene, 9,10-dibromo, 9,10-dibromo-anthracene, unii-61cp7c5y82, 9,10-dibromanthracen, 9,10-dibromo anthracenen, pubchem15115, 9,10 dibromoanthracene, anthracene,10-dibromo, 9,10-dibromoanthracene, acmc-1az4y
PubChem CID 68226
IUPAC Name 9,10-dibromoanthracene
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)Br

Specifications

Melting Point 227°C
Quantity 500 g
Formula Weight 336.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dibromoanthracene