accessibility menu, dialog, popup

UserName

CAS RN 523-27-3

IUPAC Name: 9,10-dibromoanthracene
Synonyms: 9,10-dibromoanthracene
Molecular Weight (g/mol): 336.03
Molecular Formula: C14H8Br2
InChi Key: BRUOAURMAFDGLP-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(Br)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)