Learn More
9,10-Dibromoanthracene, 98%
CAS: 523-27-3 | C14H8Br2 | 336.026 g/mol
Supplier: Thermo Scientific Chemicals L1268014
Description
9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place
Specifications
| 9,10-Dibromoanthracene | |
| 222°C to 223°C (sublimation) | |
| MFCD00001244 | |
| 1912109 | |
| anthracene, 9,10-dibromo, 9,10-dibromo-anthracene, unii-61cp7c5y82, 9,10-dibromanthracen, 9,10-dibromo anthracenen, pubchem15115, 9,10 dibromoanthracene, anthracene,10-dibromo, 9,10-dibromoanthracene, acmc-1az4y | |
| BRUOAURMAFDGLP-UHFFFAOYSA-N | |
| 9,10-dibromoanthracene | |
| 68226 | |
| 98% |
| 523-27-3 | |
| C14H8Br2 | |
| 25 g | |
| 14,3018 | |
| Soluble in water. | |
| C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)Br | |
| 336.026 | |
| 336.04 |
Chemical Identifiers
| 523-27-3 | |
| 336.026 | |
| BRUOAURMAFDGLP-UHFFFAOYSA-N | |
| 68226 | |
| C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)Br |
| C14H8Br2 | |
| MFCD00001244 | |
| anthracene, 9,10-dibromo, 9,10-dibromo-anthracene, unii-61cp7c5y82, 9,10-dibromanthracen, 9,10-dibromo anthracenen, pubchem15115, 9,10 dibromoanthracene, anthracene,10-dibromo, 9,10-dibromoanthracene, acmc-1az4y | |
| 9,10-dibromoanthracene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-342-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only