missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(diethylphosphonomethyl)anthracene 98.0+%, TCI America™

Catalog No. B28011G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28011G 1 g
B28015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28011G Supplier TCI America Supplier No. B28011G

Chemical Identifiers

CAS 60974-92-7
Molecular Formula C24H32O6P2
Molecular Weight (g/mol) 478.462
InChI Key PKLFGXZSNISEOV-UHFFFAOYSA-N
Synonym [Anthracene-9,10-diylbis(methylene)]bisphosphonic Acid Tetraethyl Ester, Tetraethyl [Anthracene-9,10-diylbis(methylene)]bisphosphonate
PubChem CID 16116648
IUPAC Name 9,10-bis(diethoxyphosphorylmethyl)anthracene
SMILES CCOP(=O)(CC1=C2C=CC=CC2=C(C3=CC=CC=C31)CP(=O)(OCC)OCC)OCC

Specifications

Melting Point 160°C
Color Yellow
Quantity 1 g
Formula Weight 478.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis(diethylphosphonomethyl)anthracene