accessibility menu, dialog, popup

UserName

H3COPOPOOCH3OCH3OCH3

CAS RN 60974-92-7

IUPAC Name: diethyl ({10-[(diethoxyphosphoryl)methyl]anthracen-9-yl}methyl)phosphonate
Synonyms: diethyl {10-[(diethoxyphosphoryl)methyl]anthracen-9-yl}methylphosphonate
Molecular Weight (g/mol): 478.46
Molecular Formula: C24H32O6P2
InChi Key: PKLFGXZSNISEOV-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=C2C=CC=CC2=C(CP(=O)(OCC)OCC)C2=CC=CC=C12)OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results