missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Bis(3,5-dihydroxyphenyl)anthracene 96.0+%, TCI America™
Supplier: TCI America B21091G
Specifications
| 9,10-Bis(3,5-dihydroxyphenyl)anthracene | |
| C26H18O4 | |
| 1 g | |
| OC1=CC(=CC(O)=C1)C1=C2C=CC=CC2=C(C2=CC(O)=CC(O)=C2)C2=CC=CC=C12 | |
| 394.43 | |
| 394.43 | |
| Crystalline Powder |
| 153715-08-3 | |
| MFCD03427350 | |
| BTBBWVMITMIXSY-UHFFFAOYSA-N | |
| 5-[10-(3,5-dihydroxyphenyl)anthracen-9-yl]benzene-1,3-diol | |
| 11047500 | |
| ≥96.0% (HPLC) |
Chemical Identifiers
| 153715-08-3 | |
| 394.43 | |
| BTBBWVMITMIXSY-UHFFFAOYSA-N | |
| 5-[10-(3,5-dihydroxyphenyl)anthracen-9-yl]benzene-1,3-diol |
| C26H18O4 | |
| MFCD03427350 | |
| 11047500 | |
| OC1=CC(=CC(O)=C1)C1=C2C=CC=CC2=C(C2=CC(O)=CC(O)=C2)C2=CC=CC=C12 |
Safety and Handling
TSCA : No