accessibility menu, dialog, popup

UserName

CAS RN 153715-08-3

IUPAC Name: 5-[10-(3,5-dihydroxyphenyl)anthracen-9-yl]benzene-1,3-diol
Synonyms: 5-[10-(3,5-dihydroxyphenyl)anthracen-9-yl]benzene-1,3-diol
Molecular Weight (g/mol): 394.43
Molecular Formula: C26H18O4
InChi Key: BTBBWVMITMIXSY-UHFFFAOYSA-N
SMILES: OC1=CC(=CC(O)=C1)C1=C2C=CC=CC2=C(C2=CC(O)=CC(O)=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results