missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(3,5-dihydroxyphenyl)anthracene 96.0+%, TCI America™

Catalog No. p-7119486
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2109100MG 100 mg
B21091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2109100MG Supplier TCI America Supplier No. B2109100MG
Only null left

Chemical Identifiers

CAS 153715-08-3
Molecular Formula C26H18O4
Molecular Weight (g/mol) 394.43
MDL Number MFCD03427350
InChI Key BTBBWVMITMIXSY-UHFFFAOYSA-N
PubChem CID 11047500
IUPAC Name 5-[10-(3,5-dihydroxyphenyl)anthracen-9-yl]benzene-1,3-diol
SMILES OC1=CC(=CC(O)=C1)C1=C2C=CC=CC2=C(C2=CC(O)=CC(O)=C2)C2=CC=CC=C12

Specifications

Quantity 100 mg
Formula Weight 394.43
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis(3,5-dihydroxyphenyl)anthracene