missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 8 mm HPLC and GC Vial Septa

Use Septa for 8-425 Screw Thread Caps to suit a variety of HPLC and GC applications

Supplier:  Thermo Scientific™ 8ST14X

 View more versions of this product

Catalog No. 13622395

Only null left
Explore available promotions

Description

Description

Choose Thermo Scientific 8 mm Septa for 8-425 Screw Thread Caps of PTFE, PTFE/Rubber or PTFE/Silicone to suit a variety of HPLC and GC applications.

  • Silicone/PTFE - Ideal for temperatures - 60 - 200°C and soft/thin septa great for thin, fragile needles.
  • PTFE - Ideal for temperatures -200 - 250°C and highest solvent stability
Specifications

Specifications

Septum for 8-425 Screw Caps
PTFE, Silicone
Non-sterile
Silicone/PTFE
50 Shore
500
Certificate of Conformance (COC)
8-425 Screw Thread Cap
8 mm
Blue
Yes
Single-use
1 pack of 500
Autoclavable
1.2 mm
Septa
8-425 Screw Thread Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.