missing translation for 'onlineSavingsMsg'
Learn More

8-Nitroquinoline 99.0+%, TCI America™

Catalog No. p-7133760
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03455G 5 g
N034525G 25 g
N0345100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N03455G Supplier TCI America Supplier No. N03455G
Only null left

Chemical Identifiers

CAS 607-35-2
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00006806
InChI Key OQHHSGRZCKGLCY-UHFFFAOYSA-N
Synonym quinoline, 8-nitro, unii-ywo6s88v4v, ccris 457, ywo6s88v4v, 8-nitroqunioline, 4msn, 8-nitro-quinoline, gilstone-yellow, zlchem 249, pubchem5825
PubChem CID 11830
IUPAC Name 8-nitroquinoline
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])N=CC=C2

Specifications

Melting Point 89°C
Color Yellow
Quantity 5 g
Formula Weight 174.16
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Nitroquinoline
RTECSNumber VC1925000
TSCA Yes