accessibility menu, dialog, popup

UserName

CAS RN 607-35-2

IUPAC Name: 8-nitroquinoline
Synonyms: 8-nitroquinoline
Molecular Weight (g/mol): 174.16
Molecular Formula: C9H6N2O2
InChi Key: OQHHSGRZCKGLCY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2N=CC=CC2=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)