missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 98%

Catalog No. aal1375103
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
AAL1375103 1 g
AAL13751MD 250 mg
2 options

Catalog No. AAL1375103

Supplier: Thermo Scientific Chemicals L1375103

Only null left

CAS: 32178-63-5 | C12H14O3 | 206.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32178-63-5
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD01075657
InChI Key VOAFGTBETRJDPU-UHFFFAOYSA-N
Synonym 8-methoxy-1,2,3,4-tetrahydronaphtalene-2-carboxylic acid, 1,2,3,4-tetrahydro-8-methoxynaphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydrophtalene-2-carboxylic acid, 2-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-8-methoxy
PubChem CID 3624709
IUPAC Name 8-methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
SMILES COC1=CC=CC2=C1CC(CC2)C(=O)O

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Beilstein 2269890
Formula Weight 206.24
Percent Purity 98%
Chemical Name or Material 8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only