accessibility menu, dialog, popup

UserName

CAS RN 32178-63-5

IUPAC Name: 8-methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
Synonyms: 8-methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: VOAFGTBETRJDPU-UHFFFAOYNA-N
SMILES: COC1=CC=CC2=C1CC(CC2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results