missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 98%

Catalog No. AAL13751MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13751MD 250 mg
AAL1375103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13751MD Supplier Thermo Scientific Chemicals Supplier No. L13751MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32178-63-5 | C12H14O3 | 206.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32178-63-5
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD01075657
InChI Key VOAFGTBETRJDPU-UHFFFAOYSA-N
Synonym 8-methoxy-1,2,3,4-tetrahydronaphtalene-2-carboxylic acid, 1,2,3,4-tetrahydro-8-methoxynaphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydrophtalene-2-carboxylic acid, 2-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-8-methoxy
PubChem CID 3624709
IUPAC Name 8-methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
SMILES COC1=CC=CC2=C1CC(CC2)C(=O)O

Specifications

Melting Point 141°C to 143°C
Quantity 250 mg
Beilstein 2269890
Formula Weight 206.24
Percent Purity 98%
Chemical Name or Material 8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only