missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 8-Hydroxy-1,3,6-pyrenetrisulfonic acid trisodium salt, 98%

Catalog No. ac412180010
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC412180010 1 g Glass bottle
AC412180050 5 g Glass bottle
2 options

Catalog No. AC412180010

Supplier: Thermo Scientific Chemicals 412180010

Only null left

CAS: 6358-69-6 | C16H7Na3O10S3 | 524.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6358-69-6
Molecular Formula C16H7Na3O10S3
Molecular Weight (g/mol) 524.37
MDL Number MFCD00037575
InChI Key KXXXUIKPSVVSAW-UHFFFAOYSA-K
PubChem CID 61388
ChEBI CHEBI:52083
IUPAC Name trisodium;8-hydroxypyrene-1,3,6-trisulfonate
SMILES [Na+].[Na+].[Na+].OC1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Green-Yellow to Yellow
Density 2.1500g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Specific Gravity 2.15
Merck Index 15, 8065
Synonym pyranine, solvent green 7, c.i. solvent green 7, japan green 204, hpts, FD&C green no. 8, japan green no. 204, pyrene 1, pyranine 120, FD&Cgreen 8
Solubility Information Solubility in water: 300g/L (25°C).
Formula Weight 524.39
Percent Purity 98%
Grade Extra Pure
Physical Form Crystals or Powder
Chemical Name or Material 8-Hydroxy-1, 3, 6-pyrenetrisulfonic acid trisodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-783-6

RUO – Research Use Only