missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-(Fmoc-amino)octanoic acid, 95%
CAS: 126631-93-4 | C23H27NO4 | 381.472 g/mol
$222.42 - $842.05
Chemical Identifiers
| CAS | 126631-93-4 |
|---|---|
| Molecular Formula | C23H27NO4 |
| Molecular Weight (g/mol) | 381.472 |
| MDL Number | MFCD00235890 |
| InChI Key | QZQXRZXYWVQWAY-UHFFFAOYSA-N |
| Synonym | n-fmoc-8-aminooctanoic acid, fmoc-8-aoc-oh, monascamine, monascorubramin, fmoc-8-aminocaprylic acid, unii-wq3ps53e2u, wq3ps53e2u, 8-9h-fluoren-9-ylmethoxycarbonylamino octanoic acid, 8-fmoc-amino-octanoic acid, 8-9h-fluoren-9-yl methoxy carbonyl amino octanoic acid |
| PubChem CID | 2756091 |
| IUPAC Name | 8-(9H-fluoren-9-ylmethoxycarbonylamino)octanoic acid |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6627903
|
Thermo Scientific Chemicals
H6627903 |
1 g |
Each for $222.42
|
|
|||||
|
AAH6627906
|
Thermo Scientific Chemicals
H6627906 |
5 g |
Each for $842.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 126631-93-4 | |
| 381.472 | |
| QZQXRZXYWVQWAY-UHFFFAOYSA-N | |
| 2756091 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCCCC(=O)O |
| C23H27NO4 | |
| MFCD00235890 | |
| n-fmoc-8-aminooctanoic acid, fmoc-8-aoc-oh, monascamine, monascorubramin, fmoc-8-aminocaprylic acid, unii-wq3ps53e2u, wq3ps53e2u, 8-9h-fluoren-9-ylmethoxycarbonylamino octanoic acid, 8-fmoc-amino-octanoic acid, 8-9h-fluoren-9-yl methoxy carbonyl amino octanoic acid | |
| 8-(9H-fluoren-9-ylmethoxycarbonylamino)octanoic acid |
Specifications
| 126631-93-4 | |
| C23H27NO4 | |
| 1 g | |
| n-fmoc-8-aminooctanoic acid, fmoc-8-aoc-oh, monascamine, monascorubramin, fmoc-8-aminocaprylic acid, unii-wq3ps53e2u, wq3ps53e2u, 8-9h-fluoren-9-ylmethoxycarbonylamino octanoic acid, 8-fmoc-amino-octanoic acid, 8-9h-fluoren-9-yl methoxy carbonyl amino octanoic acid | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCCCC(=O)O | |
| 381.472 | |
| 381.47 | |
| Powder |
| White | |
| MFCD00235890 | |
| 8286497 | |
| QZQXRZXYWVQWAY-UHFFFAOYSA-N | |
| 8-(9H-fluoren-9-ylmethoxycarbonylamino)octanoic acid | |
| 2756091 | |
| 95% | |
| 8-(Fmoc-amino)octanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only