CAS RN 126631-93-4
IUPAC Name:
8-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)octanoic acid
Synonyms:
8-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}octanoic acid
Molecular Weight (g/mol):
381.47
Molecular Formula:
C23H27NO4
InChi Key:
QZQXRZXYWVQWAY-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
8-(Fmoc-amino)octanoic acid, 95%
8-(Fmoc-amino)octanoic acid, 95%
CAS: 126631-93-4 Molecular Formula: C23H27NO4 Molecular Weight (g/mol): 381.472 MDL Number: MFCD00235890 InChI Key: QZQXRZXYWVQWAY-UHFFFAOYSA-N Synonym: n-fmoc-8-aminooctanoic acid,fmoc-8-aoc-oh,monascamine,monascorubramin,fmoc-8-aminocaprylic acid,unii-wq3ps53e2u,wq3ps53e2u,8-9h-fluoren-9-ylmethoxycarbonylamino octanoic acid,8-fmoc-amino-octanoic acid,8-9h-fluoren-9-yl methoxy carbonyl amino octanoic acid PubChem CID: 2756091 IUPAC Name: 8-(9H-fluoren-9-ylmethoxycarbonylamino)octanoic acid SMILES: C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCCCC(=O)O