missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Ethoxyquinoline-5-sulfonic Acid Hydrate 98.0+%, TCI America™
$44.28 - $121.28
Chemical Identifiers
| CAS | 15301-40-3 |
|---|---|
| Molecular Formula | C11H11NO4S |
| Molecular Weight (g/mol) | 253.27 |
| MDL Number | MFCD00065059 |
| InChI Key | YAMVZYRZAMBCED-UHFFFAOYSA-N |
| Synonym | actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol |
| PubChem CID | 23674 |
| IUPAC Name | 8-ethoxyquinoline-5-sulfonic acid |
| SMILES | CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O |
Chemical Identifiers
| 15301-40-3 | |
| 253.27 | |
| YAMVZYRZAMBCED-UHFFFAOYSA-N | |
| 23674 | |
| CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O |
| C11H11NO4S | |
| MFCD00065059 | |
| actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol | |
| 8-ethoxyquinoline-5-sulfonic acid |
Specifications
| 15301-40-3 | |
| MFCD00065059 | |
| actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol | |
| CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O | |
| 253.27 | |
| 253.27 | |
| Crystalline Powder |
| C11H11NO4S | |
| 5 g | |
| YAMVZYRZAMBCED-UHFFFAOYSA-N | |
| 8-ethoxyquinoline-5-sulfonic acid | |
| 23674 | |
| ≥98.0% (HPLC,T) | |
| 8-Ethoxyquinoline-5-sulfonic Acid Hydrate |
Safety and Handling
TSCA : No