missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Ethoxyquinoline-5-sulfonic Acid Hydrate 98.0+%, TCI America™
Supplier: TCI America E033925G
Specifications
| 8-Ethoxyquinoline-5-sulfonic Acid Hydrate | |
| C11H11NO4S | |
| 25 g | |
| YAMVZYRZAMBCED-UHFFFAOYSA-N | |
| 8-ethoxyquinoline-5-sulfonic acid | |
| 23674 | |
| ≥98.0% (HPLC,T) |
| 15301-40-3 | |
| MFCD00065059 | |
| actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol | |
| CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O | |
| 253.27 | |
| 253.27 | |
| Crystalline Powder |
Chemical Identifiers
| 15301-40-3 | |
| 253.27 | |
| YAMVZYRZAMBCED-UHFFFAOYSA-N | |
| 23674 | |
| CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O |
| C11H11NO4S | |
| MFCD00065059 | |
| actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol | |
| 8-ethoxyquinoline-5-sulfonic acid |
Safety and Handling
TSCA : No