missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 8-Cyclopentyl-1,3-dimethylxanthine
Description
8-Cyclopentyl-1,3-dimethylxanthine is a high affinity adenosine A1 receptor antagonist (pKi = 7.5-8).
Specifications
Specifications
| Chemical Name or Material | 8-Cyclopentyl-1,3-dimethylxanthine |
| CAS | 35873-49-5 |
| Synonym | 8-Cyclopentyltheophylline |
| Quantity | 100 mg |
| Solubility | Soluble to 100mM in DMSO and to 10mM in ethanol |
| Molecular Formula | C12H16N4O2 |
| Formula Weight | 248.28 |
| Physical Form | Solid |
| Color | Pale-yellow |
| Product Type | Bioactive Small Molecule |
| Show More |
For Research Use Only.