missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one 98.0+%, TCI America™

Catalog No. p-7125876
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31815G 5 g
C318125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31815G Supplier TCI America Supplier No. C31815G

Chemical Identifiers

CAS 31251-41-9
Molecular Formula C14H10ClNO
Molecular Weight (g/mol) 243.69
MDL Number MFCD00800222
InChI Key WMQNOYVVLMIZDV-UHFFFAOYSA-N
Synonym 8-Chloro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11(6H)-one
PubChem CID 9816260
IUPAC Name 8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-one
SMILES C1CC2=C(C=CC(=C2)Cl)C(=O)C3=C1C=CC=N3

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 243.69
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one
TSCA No
Recommended Storage Freezer