accessibility menu, dialog, popup

UserName

CAS RN 31251-41-9

IUPAC Name: 13-chloro-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(11),3(8),4,6,12,14-hexaen-2-one
Synonyms: 13-chloro-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(11),3(8),4,6,12,14-hexaen-2-one
Molecular Weight (g/mol): 243.69
Molecular Formula: C14H10ClNO
InChi Key: WMQNOYVVLMIZDV-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(C=C1)C(=O)C1=C(CC2)C=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results