missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 105°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 243.69 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |