missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromooctanoic Acid 97.0+%, TCI America™
Supplier: TCI America B167525G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 17696-11-6 | |
| 223.11 | |
| BKJFDZSBZWHRNH-UHFFFAOYSA-N | |
| 548275 | |
| C(CCCC(=O)O)CCCBr |
| C8H15BrO2 | |
| MFCD00004430 | |
| 8-bromocaprylic acid, 8-bromo octanoic acid, octanoic acid, 8-bromo, 8-bromo-n-octanoic acid, omega-bromocaprylic acid, 8-bromo-octanoic acid, octanoicacid, 8-bromo, acmc-1bo24, 8-bromooctanoic acid, ksc180g8h | |
| 8-bromooctanoic acid |
Specifications
| 8-Bromooctanoic Acid | |
| 37°C | |
| 150°C | |
| 25 g | |
| 3261 | |
| BKJFDZSBZWHRNH-UHFFFAOYSA-N | |
| 8-bromooctanoic acid | |
| 548275 | |
| ≥97.0% (T) |
| 17696-11-6 | |
| White-Yellow | |
| C8H15BrO2 | |
| MFCD00004430 | |
| 8-bromocaprylic acid, 8-bromo octanoic acid, octanoic acid, 8-bromo, 8-bromo-n-octanoic acid, omega-bromocaprylic acid, 8-bromo-octanoic acid, octanoicacid, 8-bromo, acmc-1bo24, 8-bromooctanoic acid, ksc180g8h | |
| C(CCCC(=O)O)CCCBr | |
| 223.11 | |
| 223.11 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No