accessibility menu, dialog, popup

UserName

OOBr

CAS RN 17696-11-6

IUPAC Name: 8-bromooctanoate
Synonyms: 8-bromooctanoate
Molecular Weight (g/mol): 222.1
Molecular Formula: C8H14BrO2
InChi Key: BKJFDZSBZWHRNH-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCCCCCCBr
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)