missing translation for 'onlineSavingsMsg'
Learn More

8-Bromooctanoic Acid 97.0+%, TCI America™

Catalog No. p-7119202
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16755G 5 g
B167525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16755G Supplier TCI America Supplier No. B16755G

Chemical Identifiers

CAS 17696-11-6
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00004430
InChI Key BKJFDZSBZWHRNH-UHFFFAOYSA-N
Synonym 8-bromocaprylic acid, 8-bromo octanoic acid, octanoic acid, 8-bromo, 8-bromo-n-octanoic acid, omega-bromocaprylic acid, 8-bromo-octanoic acid, octanoicacid, 8-bromo, acmc-1bo24, 8-bromooctanoic acid, ksc180g8h
PubChem CID 548275
IUPAC Name 8-bromooctanoic acid
SMILES C(CCCC(=O)O)CCCBr

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 150°C
Quantity 5 g
UN Number 3261
Formula Weight 223.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 8-Bromooctanoic Acid