missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoadenosine, 98%

CAS: 2946-39-6 | C10H12BrN5O4 | 346.14 g/mol

Supplier:  Thermo Scientific Chemicals 155780050

 View more versions of this product

Catalog No. AC155780050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

8-Bromoadenosine
97.5
210°C to 212°C
Authentic
Glass bottle
MFCD00005733
-54.5
VJUPMOPLUQHMLE-SBTUYZBDNA-N
(2R,3R,4S,5R)-2-(6-amino-8-bromopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
96544
346.13
Crystalline Powder
2946-39-6
100.0
White to Cream
97.5% min. (HPLC)
C10H12BrN5O4
5 g
8-bromoadenosine, bromoadenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, adenosine, 8-bromo, 6-amino-8-bromopurine riboside, 8-bromo-adenosine, 8-bromoadenine-9-beta-d-ribofuranoside, 8-bromo adenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 4s,2r,3r,5r-2-6-amino-8-bromopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol
NC1=C2N=C(Br)N([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
346.14
CHEBI:2312
98%

Chemical Identifiers

2946-39-6
346.14
VJUPMOPLUQHMLE-SBTUYZBDNA-N
96544
(2R,3R,4S,5R)-2-(6-amino-8-bromopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
C10H12BrN5O4
MFCD00005733
8-bromoadenosine, bromoadenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, adenosine, 8-bromo, 6-amino-8-bromopurine riboside, 8-bromo-adenosine, 8-bromoadenine-9-beta-d-ribofuranoside, 8-bromo adenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 4s,2r,3r,5r-2-6-amino-8-bromopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol
CHEBI:2312
NC1=C2N=C(Br)N([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 220-959-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only