accessibility menu, dialog, popup

UserName

CAS RN 2946-39-6

IUPAC Name: (2R,3R,4S,5R)-2-(6-amino-8-bromo-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: bromoadenosine
Molecular Weight (g/mol): 346.14
Molecular Formula: C10H12BrN5O4
InChi Key: VJUPMOPLUQHMLE-SBTUYZBDNA-N
SMILES: NC1=C2N=C(Br)N([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)