missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromo-1-octene 98.0+%, TCI America™
Supplier: TCI America B325725G
Specifications
| 8-Bromo-1-octene | |
| Yellow | |
| C8H15Br | |
| 25 g | |
| SNMOMUYLFLGQQS-UHFFFAOYSA-N | |
| 8-bromooct-1-ene | |
| 75907 | |
| ≥98.0% (GC) |
| 2695-48-9 | |
| 92°C | |
| MFCD00000275 | |
| 8-bromo-1-octene, 1-octene, 8-bromo, 7-octenyl bromide, 8-bromooctene, 8-azaquanine, bromo-8 octene-1, 8-bromo-oct-1-ene, acmc-1cmcf, oct-7-en-1-yl bromide | |
| C=CCCCCCCBr | |
| 191.112 | |
| 191.11 | |
| Liquid |
Chemical Identifiers
| 2695-48-9 | |
| 191.112 | |
| SNMOMUYLFLGQQS-UHFFFAOYSA-N | |
| 75907 | |
| C=CCCCCCCBr |
| C8H15Br | |
| MFCD00000275 | |
| 8-bromo-1-octene, 1-octene, 8-bromo, 7-octenyl bromide, 8-bromooctene, 8-azaquanine, bromo-8 octene-1, 8-bromo-oct-1-ene, acmc-1cmcf, oct-7-en-1-yl bromide | |
| 8-bromooct-1-ene |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : No