accessibility menu, dialog, popup

UserName

BrCH2

CAS RN 2695-48-9

IUPAC Name: 8-bromooct-1-ene
Synonyms: 8-bromooct-1-ene
Molecular Weight (g/mol): 191.11
Molecular Formula: C8H15Br
InChi Key: SNMOMUYLFLGQQS-UHFFFAOYSA-N
SMILES: BrCCCCCCC=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)