missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-1-octene 98.0+%, TCI America™

Catalog No. B32575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32575G 5 g
B325725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32575G Supplier TCI America Supplier No. B32575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2695-48-9
Molecular Formula C8H15Br
Molecular Weight (g/mol) 191.112
MDL Number MFCD00000275
InChI Key SNMOMUYLFLGQQS-UHFFFAOYSA-N
Synonym 8-bromo-1-octene, 1-octene, 8-bromo, 7-octenyl bromide, 8-bromooctene, 8-azaquanine, bromo-8 octene-1, 8-bromo-oct-1-ene, acmc-1cmcf, oct-7-en-1-yl bromide
PubChem CID 75907
IUPAC Name 8-bromooct-1-ene
SMILES C=CCCCCCCBr

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 191.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Bromo-1-octene
EINECSNumber (2)-0067
TSCA No