missing translation for 'onlineSavingsMsg'
Learn More

8-Aminoquinoline 98.0+%, TCI America™

$113.59 - $537.13

Chemical Identifiers

CAS 578-66-5
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006809
InChI Key WREVVZMUNPAPOV-UHFFFAOYSA-N
Synonym 8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
PubChem CID 11359
IUPAC Name quinolin-8-amine
SMILES C1=CC2=C(C(=C1)N)N=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A04195G
View Documents
TCI America
A04195G
5 g N/A
Only null left
 
A041925G
View Documents
TCI America
A041925G
25 g N/A
Only null left
 
A0419100G
View Documents
TCI America
A0419100G
100 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

578-66-5
144.177
WREVVZMUNPAPOV-UHFFFAOYSA-N
11359
C1=CC2=C(C(=C1)N)N=CC=C2
C9H8N2
MFCD00006809
8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
quinolin-8-amine

Specifications

578-66-5
145°C
MFCD00006809
8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
C1=CC2=C(C(=C1)N)N=CC=C2
144.177
144.18
Crystalline Powder
65°C
C9H8N2
5 g
WREVVZMUNPAPOV-UHFFFAOYSA-N
quinolin-8-amine
11359
≥98.0% (GC,T)
8-Aminoquinoline
Videos
Safety and Handling

Safety and Handling

RTECSNumber : VA9627000

TSCA : Yes

SDS
Documents

Documents