accessibility menu, dialog, popup

UserName

CAS RN 578-66-5

IUPAC Name: quinolin-8-amine
Synonyms: 8-aminoquinoline
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: WREVVZMUNPAPOV-UHFFFAOYSA-N
SMILES: NC1=C2N=CC=CC2=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (6)