missing translation for 'onlineSavingsMsg'
Learn More

8-Aminoquinoline 98.0+%, TCI America™

Catalog No. a0419100g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
A0419100G 100 g
A04195G 5 g
A041925G 25 g
3 options

Catalog No. A0419100G

Supplier: TCI America A0419100G

Only null left

Chemical Identifiers

CAS 578-66-5
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006809
InChI Key WREVVZMUNPAPOV-UHFFFAOYSA-N
Synonym 8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
PubChem CID 11359
IUPAC Name quinolin-8-amine
SMILES C1=CC2=C(C(=C1)N)N=CC=C2

Specifications

Melting Point 65°C
Boiling Point 145°C
Quantity 100 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Aminoquinoline
RTECSNumber VA9627000
TSCA Yes