missing translation for 'onlineSavingsMsg'
Learn More

8-Aminonaphthalene-2-sulfonic acid, 95%

Catalog No. p-7043055
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6530414 25 g
AAJ6530422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6530414 Supplier Thermo Scientific Chemicals Supplier No. J6530414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 119-28-8 | C10H9NO3S | 223.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-28-8
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00004032
InChI Key QEZZCWMQXHXAFG-UHFFFAOYSA-N
Synonym 8-amino-2-naphthalenesulfonic acid, 1,7-cleve's acid, 1-naphthylamine-7-sulfonic acid, 2-naphthalenesulfonic acid, 8-amino, cleve's theta-acid, 1-amino-7-sulfonaphthalene, 1-aminonaphthalene-7-sulfonic acid, 8-naphthylamine-2-sulfonic acid, 1-amino-7-naphthalenesulfonic acid, 1-amino-naphthalene-7-sulphonic acid
PubChem CID 8386
IUPAC Name 8-aminonaphthalene-2-sulfonic acid
SMILES C1=CC2=C(C=C(C=C2)S(=O)(=O)O)C(=C1)N

Specifications

Melting Point >300°C
Color Gray to Yellow
Quantity 25 g
UN Number UN2585
Beilstein 2115629
Merck Index 142,351
Formula Weight 223.25
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 8-Aminonaphthalene-2-sulfonic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
EINECSNumber 204-311-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only