accessibility menu, dialog, popup

UserName

CAS RN 119-28-8

IUPAC Name: 8-aminonaphthalene-2-sulfonic acid
Synonyms: 8-aminonaphthalene-2-sulfonic acid
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: QEZZCWMQXHXAFG-UHFFFAOYSA-N
SMILES: NC1=C2C=C(C=CC2=CC=C1)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results