missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-1-naphthalenesulfonic Acid 98.0+%, TCI America™

Supplier:  TCI America A035025G

 View more versions of this product

Catalog No. A035025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

8-Amino-1-naphthalenesulfonic Acid
Gray-Red
MFCD00035730
peri acid, 8-amino-1-naphthalenesulfonic acid, 1-naphthylamine-8-sulfonic acid, 1-naphthalenesulfonic acid, 8-amino, 8-aminonaphthalene-1-sulphonic acid, naphthylaminemonosulfonic acid s, 1-amino-8-naphthalene sulfonate, 1-aminonaphthalene-8-sulfonic acid, schollkopf's acid van, schollkopf's acid
C1=CC2=C(C(=C1)N)C(=CC=C2)S(=O)(=O)O
223.246
223.25
Crystalline Powder
82-75-7
C10H9NO3S
25 g
CYJJLCDCWVZEDZ-UHFFFAOYSA-N
8-aminonaphthalene-1-sulfonic acid
6722
≥98.0% (HPLC)

Chemical Identifiers

82-75-7
223.246
CYJJLCDCWVZEDZ-UHFFFAOYSA-N
6722
C1=CC2=C(C(=C1)N)C(=CC=C2)S(=O)(=O)O
C10H9NO3S
MFCD00035730
peri acid, 8-amino-1-naphthalenesulfonic acid, 1-naphthylamine-8-sulfonic acid, 1-naphthalenesulfonic acid, 8-amino, 8-aminonaphthalene-1-sulphonic acid, naphthylaminemonosulfonic acid s, 1-amino-8-naphthalene sulfonate, 1-aminonaphthalene-8-sulfonic acid, schollkopf's acid van, schollkopf's acid
8-aminonaphthalene-1-sulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0491

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions