missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-1-naphthalenesulfonic Acid 98.0+%, TCI America™

Catalog No. p-7116851
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A035025G 25 g
A0350100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A035025G Supplier TCI America Supplier No. A035025G

Chemical Identifiers

CAS 82-75-7
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00035730
InChI Key CYJJLCDCWVZEDZ-UHFFFAOYSA-N
Synonym peri acid, 8-amino-1-naphthalenesulfonic acid, 1-naphthylamine-8-sulfonic acid, 1-naphthalenesulfonic acid, 8-amino, 8-aminonaphthalene-1-sulphonic acid, naphthylaminemonosulfonic acid s, 1-amino-8-naphthalene sulfonate, 1-aminonaphthalene-8-sulfonic acid, schollkopf's acid van, schollkopf's acid
PubChem CID 6722
IUPAC Name 8-aminonaphthalene-1-sulfonic acid
SMILES C1=CC2=C(C(=C1)N)C(=CC=C2)S(=O)(=O)O

Specifications

Color Gray-Red
Quantity 25 g
Formula Weight 223.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 8-Amino-1-naphthalenesulfonic Acid
EINECSNumber (4)-0491
TSCA Yes