missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 700 Microliter Syringes: N Termination

The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.

Supplier:  Hamilton™ HAM80839

 View more versions of this product

Catalog No. 1481386


Only null left

Description

Description

  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications

Chemical Identifiers

Not Autoclavable
For dispensing low volumes to achieve the highest level of accuracy and precision possible
2 in.
Microliter Syringe

Specifications

Not Autoclavable
22 G
0.0161 in.
0.0283 in.
3.26 mm
0.72 mm
2 in.
51 mm
1000 psi
50°C
750
Ea.
Cone
Stainless Steel (Plunger)
0.135 in.
0.41 mm
0.305 in.
7.75 mm
For dispensing low volumes to achieve the highest level of accuracy and precision possible
2 in.
Microliter Syringe
122°F
50°F
5
500 μL
Cemented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions