missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 700 Microliter Syringes: N Termination

The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.

Supplier:  Hamilton™ HAM80384

 View more versions of this product

Catalog No. 14813090


Only null left

Description

Description

  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications

Chemical Identifiers

Not Autoclavable
For dispensing low volumes to achieve the highest level of accuracy and precision possible
2 in.
Microliter Syringe

Specifications

Not Autoclavable
26s G
0.13 mm
0.305 in.
7.75 mm
For dispensing low volumes to achieve the highest level of accuracy and precision possible
2 in.
Microliter Syringe
122°F
50°F
2
10 μL
Sharp, Bevel, Curved, Non-coring
Stainless Steel (Plunger)
0.005 in.
0.0187 in.
0.485 mm
0.47 mm
3 in.
76 mm
2000 psi
50°C
701
Ea.
5.1 cm
Cemented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions